Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
azin1b
Entrez Gene ID
317764
Gene Name
antizyme inhibitor 1b
Gene Aliases
azin1, oazin, wu:fj32d12
Chromosome Location
Chr.19: 67126 - 80685 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007159.2 NP_001007160.2 3 - 4 468 82
GenBank mRNA AB069858.1 - 1 - 2 102 82
BC061960.1 - 1 - 2 124 82
EH554389.1 - 3 - 4 507 82

View Full Product Details