Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bhmt
Entrez Gene ID
322228
Gene Name
betaine-homocysteine methyltransferase
Gene Aliases
hm:zehn2153, wu:fb53h01, wu:fb63c08, wu:fj64d01, zgc:123027
Chromosome Location
Chr.21: 40406 - 45498 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012480.1 NP_001012498.1 6 - 7 848 68
GenBank mRNA AY830415.1 - 6 - 7 848 68
BC109472.1 - 6 - 7 859 68

View Full Product Details