Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- kifbp
- Entrez Gene ID
-
100002194
- Gene Name
- kinesin family binding protein
- Gene Aliases
-
fc59b03, kbp, kiaa1279, kif1bp, unst23, wu:fc59b03, zgc:112085
- Chromosome Location
-
Chr.13: 23083856 - 23095010 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC091533.1 |
- |
2 - 3 |
571 |
134 |
View Full Product Details