Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rasl11a
Entrez Gene ID
550538
Gene Name
RAS-like, family 11, member A
Gene Aliases
zgc:110179
Chromosome Location
Chr.7: 51315634 - 51320765 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017840.2 NP_001017840.2 3 - 4 293 72
GenBank mRNA BC092775.1 - 3 - 4 446 72
BC164820.1 - 3 - 4 297 72
EF088669.1 - 3 - 4 293 72

View Full Product Details