Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
neu4
Entrez Gene ID
553569
Gene Name
sialidase 4
Gene Aliases
zgc:109998
Chromosome Location
Chr.15: 2490204 - 2497566 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020548.2 NP_001018384.1 3 - 4 539 79
GenBank mRNA BC095117.1 - 3 - 4 541 79
BC165570.1 - 2 - 3 487 79
EF116275.1 - 3 - 4 539 79

View Full Product Details