Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
klhl36
Entrez Gene ID
553604
Gene Name
kelch-like family member 36
Gene Aliases
CH73-77K24.1, zgc:110268
Chromosome Location
Chr.18: 183131 - 190743 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020580.1 NP_001018416.1 2 - 3 240 72
XM_005169934.4 XP_005169991.1 2 - 3 173 72
XM_017351924.2 XP_017207413.1 2 - 3 173 72
GenBank mRNA BC095214.1 - 2 - 3 240 72

View Full Product Details