Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
alg13
Entrez Gene ID
557181
Gene Name
ALG13 UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
otul
Chromosome Location
Chr.21: 34233496 - 34261789 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031838.1 NP_001027008.1 12 - 13 1229 58
XM_005161414.4 XP_005161471.1 12 - 13 1222 58
XM_005161417.4 XP_005161474.1 12 - 13 1222 58
GenBank mRNA AY918066.1 - 12 - 13 1229 58
BC057511.1 - 12 - 13 1108 58
BC133108.1 - 12 - 13 1136 58

View Full Product Details