Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bmp7b
Entrez Gene ID
557567
Gene Name
bone morphogenetic protein 7b
Gene Aliases
zgc:153025
Chromosome Location
Chr.23: 6421407 - 6522567 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077146.2 NP_001070614.2 4 - 5 1131 69
GenBank mRNA BC124215.1 - 4 - 5 1137 69

View Full Product Details