Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dcaf12
Entrez Gene ID
570671
Gene Name
DDB1 and CUL4 associated factor 12
Gene Aliases
wdr40a, wu:fb93d04, zgc:154031
Chromosome Location
Chr.21: 11885543 - 11911966 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077580.1 NP_001071048.1 8 - 9 1496 69
GenBank mRNA BC124796.1 - 8 - 9 1496 69

View Full Product Details