Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
d2hgdh
Entrez Gene ID
565889
Gene Name
D-2-hydroxyglutarate dehydrogenase
Gene Aliases
zgc:158661
Chromosome Location
Chr.2: 41544294 - 41562912 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080597.1 NP_001074066.1 7 - 8 1105 71
XM_005168417.4 XP_005168474.1 7 - 8 1176 71
GenBank mRNA BC129361.1 - 7 - 8 1127 71

View Full Product Details