Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
st8sia4
Entrez Gene ID
553211
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4
Gene Aliases
pst, siat 8D
Chromosome Location
Chr.10: 5667401 - 5684326 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099416.1 NP_001092886.1 4 - 5 1252 77
GenBank mRNA AJ705099.1 - 2 - 3 556 77
AJ715545.1 - 4 - 5 796 77
BC098613.1 - 4 - 5 785 77
DQ779599.1 - 4 - 5 1252 77

View Full Product Details