Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sebox
Entrez Gene ID
282666
Gene Name
SEBOX homeobox
Gene Aliases
og9x
Chromosome Location
Chr.5: 67371650 - 67382183 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320052.1 NP_001306981.1 3 - 4 469 139
GenBank mRNA AF466189.1 - 3 - 4 469 139

View Full Product Details