Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nkx3-2
Entrez Gene ID
337865
Gene Name
NK3 homeobox 2
Gene Aliases
bapx1, nkx3.2
Chromosome Location
Chr.14: 213614 - 215166 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178132.2 NP_835233.1 1 - 2 428 88
GenBank mRNA AY225416.1 - 1 - 2 428 88
BC159241.1 - 1 - 2 444 88
BC162289.1 - 1 - 2 372 88
BC162299.1 - 1 - 2 372 88

View Full Product Details