Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zfr
Entrez Gene ID
321659
Gene Name
zinc finger RNA binding protein
Gene Aliases
hnrpa1, wu:fb25b08, zgc:66235, zgc:66403
Chromosome Location
Chr.5: 189463 - 207518 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199558.2 NP_955852.2 2 - 3 162 127
GenBank mRNA BC055641.1 - - 145 127
BC059198.1 - 2 - 3 139 127

View Full Product Details