Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
coro2a
Entrez Gene ID
323538
Gene Name
coronin, actin binding protein, 2A
Gene Aliases
wu:fc02b08, zgc:73379
Chromosome Location
Chr.1: 26605029 - 26654317 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199643.2 NP_955937.1 3 - 4 480 86
XM_005159876.4 XP_005159933.1 4 - 5 715 86
XM_005159877.4 XP_005159934.1 4 - 5 736 86
GenBank mRNA BC060941.1 - 3 - 4 484 86

View Full Product Details