Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rhoq
Entrez Gene ID
327510
Gene Name
ras homolog family member Q
Gene Aliases
wu:fi15a09, zgc:55997
Chromosome Location
Chr.12: 22325 - 22796 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199818.1 NP_956112.1 3 - 4 697 83
GenBank mRNA BC045850.1 - 3 - 4 697 83
BC075783.1 - 3 - 4 698 83
BC152679.1 - 3 - 4 697 83
BC164028.1 - 3 - 4 401 83

View Full Product Details