Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- c1galt1b
- Entrez Gene ID
-
337131
- Gene Name
- core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1b
- Gene Aliases
-
c1galt1, fa98f05, wu:fa98f05, zgc:66485
- Chromosome Location
-
Chr.6: 64575 - 72708 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200051.1 |
NP_956345.1 |
3 - 4 |
1214 |
76 |
GenBank mRNA |
BC054714.1 |
- |
3 - 4 |
1214 |
76 |
View Full Product Details