Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnaja3a
Entrez Gene ID
335894
Gene Name
DnaJ heat shock protein family (Hsp40) member A3a
Gene Aliases
wu:fj36c02
Chromosome Location
Chr.3: 59528 - 81094 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201313.1 NP_958470.1 6 - 7 967 70
XM_005168471.4 XP_005168528.1 6 - 7 975 70
GenBank mRNA BC047809.1 - 6 - 7 967 70
BC165728.1 - 6 - 7 964 70

View Full Product Details