Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pak4
Entrez Gene ID
431769
Gene Name
p21 protein (Cdc42/Rac)-activated kinase 4
Gene Aliases
im:5916170, wu:fd12b01, zgc:92014
Chromosome Location
Chr.15: 25452092 - 25486421 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002222.2 NP_001002222.2 2 - 3 361 90
GenBank mRNA BC055207.1 - 2 - 3 349 90
BC074067.1 - 2 - 3 350 90

View Full Product Details