Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
immp2l
Entrez Gene ID
445299
Gene Name
inner mitochondrial membrane peptidase subunit 2
Gene Aliases
zgc:100888
Chromosome Location
Chr.25: 21298176 - 21507463 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003755.2 NP_001003755.1 2 - 3 178 70
GenBank mRNA BC078193.1 - 2 - 3 182 70

View Full Product Details