Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ric8b
Entrez Gene ID
334503
Gene Name
RIC8 guanine nucleotide exchange factor B
Gene Aliases
fj97c10, synbl, wu:fj97c10
Chromosome Location
Chr.18: 141887 - 169327 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005293.1 NP_001005293.1 1 - 2 177 128
GenBank mRNA AB354736.1 - 1 - 2 138 128
AY648760.1 - 1 - 2 177 128
BC116516.1 - 1 - 2 159 128
BC139641.1 - 1 - 2 160 128

View Full Product Details