Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fn1b
Entrez Gene ID
334613
Gene Name
fibronectin 1b
Gene Aliases
cb1057, fn1l, fn3, wu:fa14f11, wu:fb03c02
Chromosome Location
Chr.1: 5504131 - 5543086 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013261.1 NP_001013279.1 43 - 44 7136 59
XM_005160284.3 XP_005160341.1 43 - 44 7168 59
XM_009305554.3 XP_009303829.1 44 - 45 7448 59
GenBank mRNA AF500198.1 - 5 - 6 913 59
AY538257.1 - 43 - 44 7136 59
AY725818.1 - 44 - 45 7427 59

View Full Product Details