Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
myh11a
Entrez Gene ID
554168
Gene Name
myosin, heavy chain 11a, smooth muscle
Gene Aliases
myh11
Chromosome Location
Chr.6: 8499855 - 8580885 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024448.1 NP_001019619.1 23 - 24 3103 126
XM_021476875.1 XP_021332550.1 24 - 25 3156 126
XM_021476876.1 XP_021332551.1 24 - 25 3156 126
GenBank mRNA BC162152.1 - 23 - 24 3067 126
DQ022376.1 - 23 - 24 3103 126

View Full Product Details