Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
myh11a
Entrez Gene ID
554168
Gene Name
myosin, heavy chain 11a, smooth muscle
Gene Aliases
myh11
Chromosome Location
Chr.6: 8499855 - 8580885 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024448.1 NP_001019619.1 27 - 28 3757 78
XM_021476875.1 XP_021332550.1 28 - 29 3810 78
XM_021476876.1 XP_021332551.1 28 - 29 3810 78
GenBank mRNA BC162152.1 - 27 - 28 3721 78
DQ022376.1 - 27 - 28 3757 78

View Full Product Details