Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
btbd7
Entrez Gene ID
568372
Gene Name
BTB (POZ) domain containing 7
Gene Aliases
si:dkeyp-55f12.1, wu:fd08g07
Chromosome Location
Chr.20: 27113926 - 27190483 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025551.1 NP_001020722.1 6 - 7 1797 69
XM_005160586.3 XP_005160643.1 7 - 8 2366 69

View Full Product Details