Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
b4galnt1a
Entrez Gene ID
558684
Gene Name
beta-1,4-N-acetyl-galactosaminyl transferase 1a
Gene Aliases
B4GALNT1, zgc:158609
Chromosome Location
Chr.23: 28095328 - 28123841 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080570.1 NP_001074039.1 6 - 7 775 58
XM_005162256.4 XP_005162313.1 6 - 7 876 58
XM_021469816.1 XP_021325491.1 6 - 7 869 58
GenBank mRNA BC129323.1 - 6 - 7 775 58

View Full Product Details