Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
creb3l2
Entrez Gene ID
791169
Gene Name
cAMP responsive element binding protein 3-like 2
Gene Aliases
zgc:158603
Chromosome Location
Chr.4: 4710729 - 4751981 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080651.2 NP_001074120.1 4 - 5 983 118
XM_021475142.1 XP_021330817.1 3 - 4 673 118
GenBank mRNA BC129318.1 - 4 - 5 985 118

View Full Product Details