Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
drosha
Entrez Gene ID
567505
Gene Name
drosha ribonuclease III
Gene Aliases
im:7150667, rnasen, zgc:158612
Chromosome Location
Chr.12: 13344985 - 13389784 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110472.1 NP_001103942.1 22 - 23 3048 84
GenBank mRNA BC133134.1 - 22 - 23 3048 84

View Full Product Details