Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lmo2
Entrez Gene ID
30332
Gene Name
LIM domain only 2 (rhombotin-like 1)
Gene Aliases
wu:fc83a08, zgc:111930
Chromosome Location
Chr.18: 38288980 - 38296665 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131111.1 NP_571186.1 1 - 2 447 69
GenBank mRNA AF191560.1 - 1 - 2 447 69
BC093136.1 - 1 - 2 370 69

View Full Product Details