Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acvr1l
Entrez Gene ID
30615
Gene Name
activin A receptor, type 1 like
Gene Aliases
acvr1, alk2, alk8, alk8sum, etID124543.2, fk50h05, laf, lost-a-fin, mfn, wu:fk50h05
Chromosome Location
Chr.2: 41524235 - 41543760 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131345.1 NP_571420.1 6 - 7 823 72
XM_021478751.1 XP_021334426.1 5 - 6 776 72
GenBank mRNA AF038425.1 - 6 - 7 823 72
AF292028.1 - 4 - 5 635 72
BC162313.1 - 5 - 6 688 72
BC162317.1 - 5 - 6 688 72

View Full Product Details