Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
naca
Entrez Gene ID
195823
Gene Name
nascent polypeptide associated complex subunit alpha
Gene Aliases
ik:tdsubc_2b5, nkn, tdsubc_2b5, unm_t30953, xx:tdsubc_2b5
Chromosome Location
Chr.23: 17866008 - 17874341 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193352.1 NP_001180281.1 4 - 5 2705 76
NM_173264.2 NP_775371.1 3 - 4 260 76
GenBank mRNA AY099510.1 - 3 - 4 196 76
AY398384.1 - 3 - 4 200 76
BC122432.1 - 3 - 4 245 76
BC164312.1 - 2 - 3 188 76

View Full Product Details