Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fgf24
Entrez Gene ID
359831
Gene Name
fibroblast growth factor 24
Gene Aliases
SI:zC207O21.2, lbx1h, si:zc207o21.1
Chromosome Location
Chr.14: 5506984 - 5546822 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182871.3 NP_878291.2 3 - 4 865 66
GenBank mRNA AY204859.1 - 3 - 4 239 66
AY285924.1 - 3 - 4 375 66
BC121748.1 - 3 - 4 865 66
BC165417.1 - 3 - 4 273 66

View Full Product Details