Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tk1
Entrez Gene ID
327590
Gene Name
thymidine kinase 1, soluble
Gene Aliases
fi19b06, wu:fi19b06, zgc:55362
Chromosome Location
Chr.11: 45153126 - 45158056 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199832.2 NP_956126.1 4 - 5 395 124
GenBank mRNA AY192987.1 - 4 - 5 308 124
BC044148.1 - 4 - 5 380 124
BC164324.1 - 4 - 5 342 124
EH580836.1 - 4 - 5 406 124

View Full Product Details