Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ybey
Entrez Gene ID
321718
Gene Name
ybeY metalloendoribonuclease
Gene Aliases
fb30h03, wu:fb30h03, zgc:85702
Chromosome Location
Chr.9: 711822 - 715241 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328511.1 NP_001315440.1 1 - 2 374 83
NM_212594.2 NP_997759.1 1 - 2 374 83
GenBank mRNA BC067584.1 - 1 - 2 344 83
EH436338.1 - 1 - 2 398 83
EH587575.1 - 1 - 2 398 83

Gene Symbol
LOC110438491
Entrez Gene ID
110438491
Gene Name
putative ribonuclease
Gene Aliases
-
Chromosome Location
Chr.Unknown: 2436 - 6296 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021471013.1 XP_021326688.1 1 - 2 377 83
XM_021471014.1 XP_021326689.1 1 - 2 377 83

View Full Product Details