Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glmna
Entrez Gene ID
492774
Gene Name
glomulin, FKBP associated protein a
Gene Aliases
glmnl, zgc:101567
Chromosome Location
Chr.2: 10821066 - 10857900 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007415.1 NP_001007416.1 14 - 15 1549 135
XM_005171163.4 XP_005171220.1 14 - 15 1410 135
XM_021480037.1 XP_021335712.1 14 - 15 1528 135
GenBank mRNA BC085370.1 - 14 - 15 1549 135
BC165850.1 - 13 - 14 1328 135

View Full Product Details