Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zfpm2b
Entrez Gene ID
568464
Gene Name
zinc finger protein, FOG family member 2b
Gene Aliases
FOG-B, fog2b
Chromosome Location
Chr.19: 13193215 - 13286738 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039524.1 NP_001034613.1 4 - 5 688 57
XM_005159561.4 XP_005159618.1 4 - 5 682 57
GenBank mRNA AY515852.1 - 4 - 5 696 57
BC163559.1 - 4 - 5 633 57
DQ015977.1 - 4 - 5 688 57

View Full Product Details