Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kcnh2a
Entrez Gene ID
563802
Gene Name
potassium voltage-gated channel, subfamily H (eag-related), member 2a
Gene Aliases
kcnh2, kcnh2l, si:dkeyp-99c4.6
Chromosome Location
Chr.2: 24080615 - 24170737 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042722.2 NP_001036187.2 12 - 13 3432 78
XM_021480879.1 XP_021336554.1 12 - 13 3410 78
XM_021480881.1 XP_021336556.1 8 - 9 2839 78
GenBank mRNA DQ022613.1 - 12 - 13 2972 78
HM209079.1 - 12 - 13 2972 78

View Full Product Details