Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
amotl2a
Entrez Gene ID
558920
Gene Name
angiomotin like 2a
Gene Aliases
amotl2, cb308, sb:cb308, wu:fc41b07, wu:fc64c06
Chromosome Location
Chr.6: 27992017 - 27999374 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080177.1 NP_001073646.1 8 - 9 1964 68
GenBank mRNA BC163299.1 - 8 - 9 1838 68
DQ887096.1 - 8 - 9 1964 68

View Full Product Details