Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
olfcu1
Entrez Gene ID
794431
Gene Name
olfactory receptor C family, u1
Gene Aliases
si:dkey-183j10.1, v2rx1
Chromosome Location
Chr.18: 165447 - 169730 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082899.1 NP_001076368.1 5 - 6 1703 77

View Full Product Details