Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
terb1
Entrez Gene ID
559140
Gene Name
telomere repeat binding bouquet formation protein 1
Gene Aliases
ccdc79, si:dkey-30c15.5
Chromosome Location
Chr.18: 104300 - 123180 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089382.1 NP_001082851.1 15 - 16 2119 75

View Full Product Details