Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cbln4
Entrez Gene ID
563847
Gene Name
cerebellin 4 precursor
Gene Aliases
zgc:172190
Chromosome Location
Chr.23: 37955041 - 37974323 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110391.1 NP_001103861.1 2 - 3 797 103
GenBank mRNA BC154577.1 - 2 - 3 797 103

View Full Product Details