Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tsr2
Entrez Gene ID
317639
Gene Name
TSR2 ribosome maturation factor
Gene Aliases
chunp6926, dt1p1a10l, zgc:109784
Chromosome Location
Chr.8: 8341082 - 8349665 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173284.3 NP_775391.1 2 - 3 223 86
GenBank mRNA AY099514.1 - 1 - 2 119 86
AY398337.1 - 2 - 3 203 86
BC066714.1 - 2 - 3 203 86
BC078277.1 - 2 - 3 227 86
BC092688.1 - 2 - 3 239 86
BC165180.1 - 1 - 2 121 86

View Full Product Details