Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tshba
Entrez Gene ID
353223
Gene Name
thyroid stimulating hormone subunit beta a
Gene Aliases
tshb
Chromosome Location
Chr.6: 182019 - 186128 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181494.2 NP_852471.1 2 - 3 208 69
XM_021476792.1 XP_021332467.1 2 - 3 379 69
XM_021476793.1 XP_021332468.1 2 - 3 306 69
GenBank mRNA AY135147.1 - 2 - 3 208 69
BC163604.1 - 1 - 2 201 69
BC163605.1 - 1 - 2 201 69

View Full Product Details