Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dcun1d1
Entrez Gene ID
327232
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae)
Gene Aliases
fd19a01, wu:fd19a01, zgc:66414
Chromosome Location
Chr.2: 7698446 - 7711498 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199772.1 NP_956066.1 2 - 3 263 104
GenBank mRNA BC057530.1 - 2 - 3 263 104
BC068381.1 - 2 - 3 229 104

View Full Product Details