Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
anp32a
Entrez Gene ID
335907
Gene Name
acidic (leucine-rich) nuclear phosphoprotein 32 family, member A
Gene Aliases
wu:fj36e08, zgc:55516
Chromosome Location
Chr.7: 33345627 - 33350002 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199986.1 NP_956280.1 3 - 4 413 97
GenBank mRNA BC048047.1 - 3 - 4 413 97
BC071357.1 - 3 - 4 468 97
BC165448.1 - 3 - 4 360 97

View Full Product Details