Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdcd4a
Entrez Gene ID
406261
Gene Name
programmed cell death 4a
Gene Aliases
wu:fa55g03, wu:fb08a11, wu:fe01a02, zgc:66311
Chromosome Location
Chr.1: 26046146 - 26063196 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212988.1 NP_998153.1 6 - 7 889 93
XM_005159885.4 XP_005159942.1 7 - 8 1130 93
GenBank mRNA BC054680.1 - 6 - 7 889 93

View Full Product Details