Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ccng2
Entrez Gene ID
266794
Gene Name
cyclin G2
Gene Aliases
cb438, wu:fa08f07, wu:fk28c06, wu:fk91a03, zgc:77125
Chromosome Location
Chr.21: 5800354 - 5807559 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213172.1 NP_998337.1 5 - 6 636 79
GenBank mRNA BC066368.1 - 5 - 6 636 79

View Full Product Details