Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crip2
Entrez Gene ID
554826
Gene Name
cysteine-rich protein 2
Gene Aliases
wu:fb30h10, zgc:55408
Chromosome Location
Chr.17: 37445525 - 37474633 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213497.2 NP_998662.2 3 - 4 282 73
GenBank mRNA BC044391.1 - 3 - 4 282 73
BC159242.1 - 3 - 4 272 73
BC164298.1 - 3 - 4 232 73

View Full Product Details