Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ankrd9
Entrez Gene ID
492328
Gene Name
ankyrin repeat domain 9
Gene Aliases
zgc:92109
Chromosome Location
Chr.17: 29276559 - 29277731 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007294.1 NP_001007295.1 1 - 1 1052 136
GenBank mRNA BC085635.1 - 1 - 1 1052 136

View Full Product Details