Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mffa
Entrez Gene ID
393508
Gene Name
mitochondrial fission factor a
Gene Aliases
mff, zgc:66022
Chromosome Location
Chr.15: 35226673 - 35252559 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200536.1 NP_956830.1 6 - 6 1451 86
XM_005157755.4 XP_005157812.1 8 - 8 1725 86
XM_005157756.4 XP_005157813.1 7 - 7 1559 86
XM_021481359.1 XP_021337034.1 7 - 7 1511 86
GenBank mRNA BC056301.1 - 6 - 6 1451 86

View Full Product Details